Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Charts
Card Sheet
Card Sheet
Card Sheet
Catalog # MUEL19.068.05
Supplier:
MUELLER KREMPEL
Expand image
Card Sheet
Catalog # MUEL19.068.05
Supplier:
MUELLER KREMPEL
Specifications
Colour:
Yellow
Description:
Map sheets
Pk:
60
W×H:
65×40 mm
Specifications
About this item
Product Details
This card sheet is made of celluloid.
Product Details