KORASILON™ pastes
Supplier: Merck
Certificates
About this item
The pastes are excellently suitable as glide-, lubricant- and release agent for e. g. vales, joints and gaskets, ground joints, cables and wires. They are frequently also used as assembly aid as well as protection- and sealing aid. Furthermore, we recommend the pastes for maintaining rubber seals. In electronics, they find their use for the assembly of insulators and switchgears amongst others.
- Stable at temperatures ranging from -50 °C to more than 300 °C
- Exceptional hydrophobic and practically insoluble in water
- Show thixotropic behavior
Frequently Bought Together