Order Entry
Austria
ContactUsLinkComponent
Racks for test tubes, stainless steel
Racks for test tubes, stainless steel
Catalog # WCDF01246
Racks for test tubes, stainless steel
Catalog # WCDF01246

Specifications

  • Hole Ø:
    18 mm
  • No. of Holes:
    48 (4×12)
  • Height:
    70 mm
  • L×W:
    287×101 mm
  • Material:
    Stainless steel
  • Pk:
    1

Specifications

About this item

Racks made of stainless steel wire.

  • Very good chemical resistance
  • Excellent thermal resistance
  • Robust, stable construction and easy to clean