ContactUsLinkComponent
Immersion oil
Immersion oil
Catalog # APPCA0699.0100
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Immersion oil
Catalog # APPCA0699.0100
Supplier:  PanReac AppliChem
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    Immersion oil
  • Seq:
    01
Specifications
About this item
This fluorescence-free immersion oil for microscopy is highly UV-permeable and free of PCB's.