ContactUsLinkComponent
Microfiltration assembly, ULTRA-WARE®, KIMBLE®
Microfiltration assembly, ULTRA-WARE®, KIMBLE®
Catalog # SCERSP179950-4532
Supplier:  DWK Life Sciences
undefined
Microfiltration assembly, ULTRA-WARE®, KIMBLE®
Catalog # SCERSP179950-4532
Supplier:  DWK Life Sciences
Specifications
  • Description:
    Microfiltration assembly with GL 45 thread
  • Filters:
    47 - 90 mm
  • Pk:
    1
Specifications
About this item
This microfiltration adapter assembly is designed to allow connection of a 47 or 90 mm filtration support base with a straight drip tip to a bottle or reservoir with a GL 45 thread.
Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly.
Certifications: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements