ContactUsLinkComponent
High content cell imaging system, ImageXpress Micro 4
High content cell imaging system, ImageXpress Micro 4
Supplier:  Molecular Devices
Certificates
undefined
High content cell imaging system, ImageXpress Micro 4
Supplier:  Molecular Devices
Specifications
  • Catalog No:
  • MLDV5039859
  • MLDV5039857
  • MLDV5039856
  • MLDV5037556
  • MLDV5039858
  • MLDVMICRO4
  • Magnification:
  • 1 – 100×
  • Condenser:
  • 65 mm WD and PhL, Ph1, and Ph2 selectable phase rings
  • Illumination:
  • Broad spectrum light source − Optional upgrade
  • Sensor:
  • High quantum efficiency, 16 bit CMOS
  • Software:
  • MetaXpress including application module, custom module, and journal
  • W×D×H:
  • 530×860×480 mm
  • Weight:
  • 95 kg
Specifications
Objectives
Product Information
Frequently Bought Together