Electrolysis apparatus according to Hoffmann
Supplier: NEUBERT VOLUME GLASSWAERE
Certificates
About this item
Electrolysis apparatus according to Hoffmann, made of borosilicate glass 3.3.
- Connecting piece for more stability
- Glass tap NS 12
- Graduated from 0 - 60 ml with 0,2 ml intervals
Frequently Bought Together