Order Entry
Austria
ContactUsLinkComponent
Cleaning concentrate, TICKOPUR TR 13
Cleaning concentrate, TICKOPUR TR 13
Catalog # 142-1991
undefined
Cleaning concentrate, TICKOPUR TR 13
Catalog # 142-1991

Specifications

  • Description:
    TICKOPUR TR 13
  • Pk:
    5 L
  • pH value:
    11.9 (1%)

Specifications

About this item

TICKOPUR TR 13 is a intensive cleaner used for the removal of stubborn dirt.

  • Dosage: 5%
  • Alkaline, pH 11,9 (1%)
  • Without phosphate and silicate
Caution: Alkali sensitive parts like light metals, tin, zinc and non-ferrous metals may be damaged.