Order Entry
Austria
ContactUsLinkComponent
 
Cleaning cloths, GP Extra
  VILE115090
undefined
Cleaning cloths, GP Extra
  VILE115090

 

  • Packed:
    25/pack
  • Colour:
    Yellow
  • L×W:
    290×365 mm
  • Seq:
    04
  • Pk:
    25

 

 

Economical cloths with non woven structure for general purpose wet and damp cleaning tasks.

  • Non woven structure: Growth of bacteria and germs is greatly reduced
  • Easily lifts dirt from surfaces
  • High fibre density greatly improves absorbency
  • Easy to rinse
  • Washable at 95 °C