ContactUsLinkComponent
Ion exchange resins, Diaion® PA312 (Cl)
Ion exchange resins, Diaion® PA312 (Cl)
Catalog # 46426.A1
CAS Number:  
Ion exchange resins, Diaion® PA312 (Cl)
Catalog # 46426.A1
CAS Number:  
Specifications
  • Particle Size:
    300 − 1180 µm
  • Pk:
    1 kg
  • Description:
    Ion exchange resin, 6% cross-linked, strongly basic porous type, 1,2 meq/ml on PS-DVB
Specifications
About this item
These resins are ideal for use in decolourisation in sugar solutions, metal recovery, pharmaceuticals and purification of amino acids.