ContactUsLinkComponent
Handheld refractometer, HHR-2N
Handheld refractometer, HHR-2N
Catalog # 635-0140
Supplier:  Atago
Handheld refractometer, HHR-2N
Catalog # 635-0140
Supplier:  Atago
Specifications
  • Model:
    HHR-2N
  • Range:
    12 − 30 % Bx
  • Resolution:
    0,1 %
  • Description:
    Handheld refractometer (for honey)
  • Pk:
    1
Specifications
About this item
This refractometer is designed to determine the water content in honey.
  • Scale to measure the percentage of water
  • Refractive index (RI) of sucrose
  • Determination of moisture in honey
Specifically suitable for use in food applications.