ContactUsLinkComponent
Square cover glasses
Square cover glasses
Catalog # HECH92101401240
Square cover glasses
Catalog # HECH92101401240
Specifications
  • Description:
    Square cover glasses
  • L×W:
    30×30 mm
  • Thickness:
    0,17 ±0,01 mm
  • Pk:
    1.000
Specifications
About this item
Square cover glasses are made up of colourless borosilicate glass.
  • Low levels of inclusions, striae, bubbles and streaks
  • High spectral transmission, excellent flatness, optimum resistance to chemicals
  • Refractive index finely adapted to microscopes
Certifications: ISO 8255