Order Entry
Austria
ContactUsLinkComponent
Multi-parameter meters, handheld, SevenGo Duo™ / Duo pro™ (SG23 / SG78)
Multi-parameter meters, handheld, SevenGo Duo™ / Duo pro™ (SG23 / SG78)
Supplier:  Mettler - Toledo
Certificates
undefined
Multi-parameter meters, handheld, SevenGo Duo™ / Duo pro™ (SG23 / SG78)
Supplier:  Mettler - Toledo

Specifications

  • Catalog No:
  • 664-0163
  • 665-0192
  • 665-0201
  • 664-0161
  • 665-0191
  • 665-0200
  • 665-0195
  • 664-0162
  • 665-0196
  • Model:
  • SG23
  • SG23
  • SG78
  • SG78
  • SG23
  • SG23
  • SG78
  • SG78
  • SG78
  • pH range:
  • 0,00...+14,00
  • 0,00...+14,00
  • –2,000...+19,999
  • –2,000...+19,999
  • 0,00...+14,00
  • 0,00...+14,00
  • –2,000...+19,999
  • –2,000...+19,999
  • –2,000...+19,999
  • pH resolution:
  • 0,01
  • 0,01
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,01
  • 0,01
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • pH accuracy:
  • ±0,01
  • ±0,01
  • ±0,002
  • ±0,002
  • ±0,01
  • ±0,01
  • ±0,002
  • ±0,002
  • ±0,002
  • pH calibration:
  • Max. 3 points, 4 pre-defined buffer groups
  • Max. 3 points, 4 pre-defined buffer groups
  • Max. 5 points, 7 pre-defined buffer groups
  • Max. 5 points, 7 pre-defined buffer groups
  • Max. 3 points, 4 pre-defined buffer groups
  • Max. 3 points, 4 pre-defined buffer groups
  • Max. 5 points, 7 pre-defined buffer groups
  • Max. 5 points, 7 pre-defined buffer groups
  • Max. 5 points, 7 pre-defined buffer groups
  • mV range:
  • ±1999
  • ±1999
  • ±1999
  • ±1999
  • ±1999
  • ±1999
  • ±1999
  • ±1999
  • ±1999
  • mV resolution:
  • 1
  • 1
  • 0,1
  • 0,1
  • 1
  • 1
  • 0,1
  • 0,1
  • 0,1
  • mV accuracy:
  • ±1
  • ±1
  • ±0,1
  • ±0,1
  • ±1
  • ±1
  • ±0,1
  • ±0,1
  • ±0,1
  • Conductivity range:
  • 0,01 µS/cm - 500 mS/cm
  • 0,01 µS/cm - 500 mS/cm
  • 0,01 µS/cm - 1000 mS/cm
  • 0,01 µS/cm - 1000 mS/cm
  • 0,01 µS/cm - 500 mS/cm
  • 0,01 µS/cm - 500 mS/cm
  • 0,01 µS/cm - 1000 mS/cm
  • 0,01 µS/cm - 1000 mS/cm
  • 0,01 µS/cm - 1000 mS/cm
  • Conductivity resolution:
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • Conductivity accuracy:
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • Resistivity range:
  • 0,00 - 100,00 MΩ/cm
  • 0,00 - 100,00 MΩ/cm
  • 0,00 - 100,00 MΩ/cm
  • 0,00 - 100,00 MΩ/cm
  • 0,00 - 100,00 MΩ/cm
  • 0,00 - 100,00 MΩ/cm
  • 0,00 - 100,00 MΩ/cm
  • 0,00 - 100,00 MΩ/cm
  • 0,00 - 100,00 MΩ/cm
  • Salinity range:
  • 0,00 - 80,00 ppt
  • 0,00 - 80,00 ppt
  • 0,00 - 80,00 ppt
  • 0,00 - 80,00 ppt
  • 0,00 - 80,00 ppt
  • 0,00 - 80,00 ppt
  • 0,00 - 80,00 ppt
  • 0,00 - 80,00 ppt
  • 0,00 - 80,00 ppt
  • TDS range:
  • 0,01 mg/l - 300 g/l
  • 0,01 mg/l - 300 g/l
  • 0,01 mg/l - 600 g/l
  • 0,01 mg/l - 600 g/l
  • 0,01 mg/l - 300 g/l
  • 0,01 mg/l - 300 g/l
  • 0,01 mg/l - 600 g/l
  • 0,01 mg/l - 600 g/l
  • 0,01 mg/l - 600 g/l
  • TDS resolution:
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • 0,01 / 1
  • TDS accuracy:
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • Temperature range:
  • –5,0...+105,0 °C
  • –5,0...+105,0 °C
  • pH/mV: ATC: –5,0...+105,0; MTC: –30...+130
    Cond: –5,0...+105,0 °C
  • pH/mV: ATC: –5,0...+105,0; MTC: –30...+130
    Cond: –5,0...+105,0 °C
  • –5,0...+105,0 °C
  • –5,0...+105,0 °C
  • pH/mV: ATC: –5,0...+105,0; MTC: –30...+130
    Cond: –5,0...+105,0 °C
  • pH/mV: ATC: –5,0...+105,0; MTC: –30...+130
    Cond: –5,0...+105,0 °C
  • pH/mV: ATC: –5,0...+105,0; MTC: –30...+130
    Cond: –5,0...+105,0 °C
  • Temperature resolution:
  • pH/mV: 0,1
    Cond: 0,1 °C
  • pH/mV: 0,1
    Cond: 0,1 °C
  • pH/mV: 0,1
    Cond: 0,1 °C
  • pH/mV: 0,1
    Cond: 0,1 °C
  • pH/mV: 0,1
    Cond: 0,1 °C
  • pH/mV: 0,1
    Cond: 0,1 °C
  • pH/mV: 0,1
    Cond: 0,1 °C
  • pH/mV: 0,1
    Cond: 0,1 °C
  • pH/mV: 0,1
    Cond: 0,1 °C
  • Temperature accuracy:
  • pH/mV: ±0,5 °C
    Cond: ±0,2 °C
  • pH/mV: ±0,5 °C
    Cond: ±0,2 °C
  • pH/mV: ±0,2 °C
    Cond: ±0,1 °C
  • pH/mV: ±0,2 °C
    Cond: ±0,1 °C
  • pH/mV: ±0,5 °C
    Cond: ±0,2 °C
  • pH/mV: ±0,5 °C
    Cond: ±0,2 °C
  • pH/mV: ±0,2 °C
    Cond: ±0,1 °C
  • pH/mV: ±0,2 °C
    Cond: ±0,1 °C
  • pH/mV: ±0,2 °C
    Cond: ±0,1 °C
  • Temperature compensation:
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Display:
  • High contrast LCD
  • High contrast LCD
  • Backlit, dot matrix LCD
  • Backlit, dot matrix LCD
  • High contrast LCD
  • High contrast LCD
  • Backlit, dot matrix LCD
  • Backlit, dot matrix LCD
  • Backlit, dot matrix LCD
  • Power supply:
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • 4 AA batteries 1,5 V or NiMH accumulator 1,3 V
  • Compliance:
  • GLP / IP 67
  • GLP / IP 67
  • GLP / IP 67
  • GLP / IP 67
  • GLP / IP 67
  • GLP / IP 67
  • GLP / IP 67
  • GLP / IP 67
  • GLP / IP 67
  • Data logging capacity:
  • 99 data records
  • 99 data records
  • 500 data records
  • 500 data records
  • 99 data records
  • 99 data records
  • 500 data records
  • 500 data records
  • 500 data records
  • Connections:
  • pH: BNC/RCA (NTC 30 kOhm)
    Cond: LTW 7-pin
  • pH: BNC/RCA (NTC 30 kOhm)
    Cond: LTW 7-pin
  • pH: BNC/RCA (NTC 30 kOhm)
    Cond: LTW 7-pin
  • pH: BNC/RCA (NTC 30 kOhm)
    Cond: LTW 7-pin
  • pH: BNC/RCA (NTC 30 kOhm)
    Cond: LTW 7-pin
  • pH: BNC/RCA (NTC 30 kOhm)
    Cond: LTW 7-pin
  • pH: BNC/RCA (NTC 30 kOhm)
    Cond: LTW 7-pin
  • pH: BNC/RCA (NTC 30 kOhm)
    Cond: LTW 7-pin
  • pH: BNC/RCA (NTC 30 kOhm)
    Cond: LTW 7-pin
  • Outputs:
  • -
  • -
  • IR to printer or PC via RS232 or USB adapter
  • IR to printer or PC via RS232 or USB adapter
  • -
  • -
  • IR to printer or PC via RS232 or USB adapter
  • IR to printer or PC via RS232 or USB adapter
  • IR to printer or PC via RS232 or USB adapter
  • W×D×H:
  • 90×45×220 mm
  • 90×45×220 mm
  • 90×45×220 mm
  • 90×45×220 mm
  • 90×45×220 mm
  • 90×45×220 mm
  • 90×45×220 mm
  • 90×45×220 mm
  • 90×45×220 mm
  • Weight:
  • 0,357 kg (without batteries)
  • 0,357 kg (without batteries)
  • 0,325 kg (without batteries)
  • 0,325 kg (without batteries)
  • 0,357 kg (without batteries)
  • 0,357 kg (without batteries)
  • 0,325 kg (without batteries)
  • 0,325 kg (without batteries)
  • 0,325 kg (without batteries)

Specifications

Related Information

Electrodes

Product Information

Frequently Bought Together