Cellulose for preparative chromatography
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Micro-crystalline cellulose is a hydrophilic polysaccharide packing material preferred for the raw separation of amino acids and related compounds. Cellulose is also used for the gentle purification of bio-molecules. Because of the organic nature of cellulose, it can only be used in pre-swollen state and under gentle or hydrostatic pressure.
Frequently Bought Together