ContactUsLinkComponent
DNA fingerprinting kit
DNA fingerprinting kit
  BE-104
undefined
DNA fingerprinting kit
  BE-104
  • Description:
    DNA fingerprinting
  • Environmentally Preferable:
  • Pk:
    1 KIT
This DNA fingerprinting kit allows students to carry out their own criminal investigation by comparing DNA samples collected from suspects to DNA collected at a pseudo-crime scene.
  • Introduces DNA fingerprinting technique
  • Teaches DNA fragmentation by specific enzymes
  • Introduction to agarose electrophoresis
Students will use specialised molecules to cut the DNA into fragments, which can then be visualised by a process known as electrophoresis. The resulting pattern, or fingerprint, can be compared and the guilty suspect identified.
 : Supplied with components needed for hands-on experimentation for six groups of 4 - 5 students or 24 - 30 students with teacher’s guide and separate student’s guides.