Order Entry
Austria
ContactUsLinkComponent
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution N1 indicator
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution N1 indicator
Catalog # LOVI56L017165
Supplier:  Lovibond
CAS Number:  
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution N1 indicator
Catalog # LOVI56L017165
Supplier:  Lovibond
CAS Number:  

Specifications

  • Pack type:
    Dropping bottle
  • Pk:
    65 mL
  • Seq:
    2390

Specifications

About this item

 :
 :692.54 g/mol
CAS Number:14634-91-4
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428