ContactUsLinkComponent
Stopcock, three way, BD Connecta™
Stopcock, three way, BD Connecta™
Catalog # BDAM394501
Supplier:  BD Medical
Stopcock, three way, BD Connecta™
Catalog # BDAM394501
Supplier:  BD Medical
Specifications
  • Description:
    Stopcock, three way, with BD Q-Syte™ luer access split septum device
  • Pk:
    50
Specifications
About this item
This three way stopcock has a flow rate of 23,4 L/hr and a prime volume of 0,31 ml.