Order Entry
Austria
ContactUsLinkComponent
Must meter
Must meter
Catalog # ALLA2000FG10/20QP
Supplier:  Alla France
undefined
Must meter
Catalog # ALLA2000FG10/20QP
Supplier:  Alla France

Specifications

  • Description:
    Must meter
  • Division:
    0,001 g/ml ; 0,5 % Vol
  • Pk:
    1
  • Range:
    0,990 – 1,120 g/ml ; 3 – 17 % Vol

Specifications

About this item

Dual scale hydrometer designed for use with wine, beer, and cider.

  • Calibration temperature of 20 °C

Suitable for measurements in must.