ContactUsLinkComponent
VWR®, Turbidity standards Set
VWR®, Turbidity standards Set
Catalog # 84209.600
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Turbidity standards Set
Catalog # 84209.600
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    4 sealed cells
  • Pk:
    1 KIT
  • Turbidity:
    200 NTU
    800 NTU
    20 NTU
    < 0,1 NTU
  • Description:
    Set of turbidity standards TSK (< 0,1; 20; 200; 800 NTU)
Specifications
About this item
Turbidity standards in kit form or alternatively as formazine stock solutions which allows users to create their own defined standards.
  • Set of standards designed for use with turbidity meters TIR 200 and TIR 210 with a measuring range from 0,01 to 1100 NTU
  • Ideal for units that measure scattered light at an angle of 90 0, as stipulated in EN ISO 7027
Certifications: ISO 7027
Delivery: Kit supplied in handy case