ContactUsLinkComponent
Swabs in tubes, Dryswab™
Swabs in tubes, Dryswab™
Catalog # MEDWMW1028D
Swabs in tubes, Dryswab™
Catalog # MEDWMW1028D
Specifications
  • Sterile:
    +
  • Description:
    Swabs in tubes, Dryswab™
  • Length:
    150 mm
  • Material:
    Polyester
  • Seq:
    02
  • Cap colour:
    Black
  • Pk:
    800
Specifications
About this item
Individual labelled sterile tubes with polyester bud, standard plastic shaft.
  • Tamper-evident label is clearly printed
  • Lot number and expiry date included
Recommended for clinical use, forensics, as well as environmental tests.
Shelf life of five years.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904