Order Entry
Austria
ContactUsLinkComponent
 
pH indicator, universal reagents
Catalog # LOVI451771
Supplier:  Lovibond
CAS Number:  
undefined
pH indicator, universal reagents
Catalog # LOVI451771
Supplier:  Lovibond
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Universal solution, liquid reagent, 100 ml
  • Range:
    4,0 – 11 pH

Specifications

About this item

Indicator for pH range 4,0 to 11.

Can be used for measuring pH in conjunction with the Comparator 2000+ system.(disc 2/1P)