pH indicator, bromo thymol bluel tablets, Lovibond®
Supplier: Lovibond
Certificates
About this item
Indicator for pH range 6,0 to 7,6.
Can be used for measuring pH in conjunction with the Comparator 2000+ system.
Frequently Bought Together
Indicator for pH range 6,0 to 7,6.
Can be used for measuring pH in conjunction with the Comparator 2000+ system.