Order Entry
Austria
ContactUsLinkComponent
PHMB tablet for use with Lovibond® MD photometers
PHMB tablet for use with Lovibond® MD photometers
Catalog # LOVI516100BT
Supplier:  Lovibond
CAS Number:  
undefined
PHMB tablet for use with Lovibond® MD photometers
Catalog # LOVI516100BT
Supplier:  Lovibond
CAS Number:  

Specifications

  • Chemical method:
    Buffer/indicator
  • Pk:
    100 Tab.
  • Description:
    PHMB photometer tablet
  • For unit:
    MD600, MD610
  • Range:
    2 - 60 mg/l PHMB

Specifications

About this item