ContactUsLinkComponent
Charging pipette carousel for four pipettes
Charging pipette carousel for four pipettes
Artikelnr. 613-4124
Supplier:  Sartorius
   Low Manufacturing Impact
Charging pipette carousel for four pipettes
Artikelnr. 613-4124
Supplier:  Sartorius
   Low Manufacturing Impact
Specifications
  • Colour:
    White
  • Description:
    Charging carousel for 4 pipettes
  • Seq:
    64
  • W×D×H:
    380×145×115 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    Renewable energy certificate; ISO 14001
  • Pk:
    1
Specifications
About this item
The charging carousel for four pipettes is a convenient, easy-to-use storage and charge system designed to keep your electronic pipettes ready for use and safe from contamination.
  • Charging carousel has a simple, elegant design
  • Holds pipettes in an upright position while not in use to avoid contamination from benches and tables
  • Compact carousel stand: Saves valuable bench space
Proper use and storage will ensure long-lasting use of your electronic pipettes, keeping them ready for whenever you need them. When storing your electronic pipette (or any pipette), it should be stored in an upright position within the stand. Storing in this manner helps ensure cleanliness and safety while avoiding contamination from things like desks and benches. It is recommended that electronic pipettes be stored and charged on a charging carousel whenever they are not in use. In this way, the batteries always remain charged, ready for use whenever work begins. The Charging carousel was designed with compact storage needs in mind.
As more vital technology and products become available for improving workflows, streamlining applications and keeping up with demand, it is vital to ensure your workspace is efficiently organised. These compact carousels are ideal for saving bench space and ensuring a safe and clean environment. Regardless of the pipette (e.g., channel number) being used, the Charging Carousel can help usage of your Picus® electronic pipettes.