About this item
Superspher® improves the separation efficiency of HPLC analysis. This high performance spherical silica carrier, with a mean particle size of 4 μm, yields the best pressure/separation performance ratio in view of today's generation of HPLC systems and according to theoretical calculation and practical experience.
The number of theoretical plates for Superspher® is <100000 N/m. Therefore, they are always first choice if complex mixtures demand high peak capacity.
A broad range of modifications on Superspher® are available: Non polar derivatives (RP-8, RP-8 endcapped, RP-18, RP-18 endcapped and RP-select B) and polar derivatives (Si 60).
Guard cartridges have to be connected to the LiChroCART® cartridges by using manu-CART® cartridge holder 1.51486.0001
27 Options Available Below
Product Details & Documents
Superspher® improves the separation efficiency of HPLC analysis. This high performance spherical silica carrier, with a mean particle size of 4 μm, yields the best pressure/separation performance ratio in view of today's generation of HPLC systems and according to theoretical calculation and practical experience.
The number of theoretical plates for Superspher® is <100000 N/m. Therefore, they are always first choice if complex mixtures demand high peak capacity.
A broad range of modifications on Superspher® are available: Non polar derivatives (RP-8, RP-8 endcapped, RP-18, RP-18 endcapped and RP-select B) and polar derivatives (Si 60).
Guard cartridges have to be connected to the LiChroCART® cartridges by using manu-CART® cartridge holder 1.51486.0001
Documents
Product Family Options
Product Information
- PhaseParticle SizeØ int.LengthAvailabilityPrice
- 1.16869.0001Superspher® 100 RP-18 endcapped LiChroCART®Superspher® 100 RP-18 endcapped LiChroCART®RP-184 µm4,00 mm25 mmLog in for availability1 * 3 ST769,00 €
- 1.16855.0001Superspher® 100 RP-18 endcapped LiChroCART®Superspher® 100 RP-18 endcapped LiChroCART®RP-184 µm4,00 mm125 mmLog in for availability1 * 1 ST883,00 €
- 1.16858.0001Superspher® 100 RP-18 endcapped LiChroCART®Superspher® 100 RP-18 endcapped LiChroCART®RP-184 µm4,00 mm250 mmLog in for availability1 * 1 ST993,00 €
- 1.50792.0001Superspher® 100 RP-18 LiChroCART®
- 1.51299.0001Superspher® 100 RP-18 LiChroCART®
- 1.16051.0001Superspher® 100 RP-18 LiChroCART®
- 1.50980.0001Superspher® 100 RP-18 LiChroCART®
- 1.16056.0001Superspher® 100 RP-18 LiChroCART®
- 1.51299.1003Superspher® 100 RP-18 LiChroCART® (3 columns from 3 batches)Superspher® 100 RP-18 LiChroCART® (3 columns from 3 batches)RP-184 µm3,00 mm250 mmLog in for availability1 * 1 KIT
- 1.16051.1003Superspher® 100 RP-18 LiChroCART® (3 columns from 3 batches)Superspher® 100 RP-18 LiChroCART® (3 columns from 3 batches)RP-184 µm4,00 mm125 mmLog in for availability1 * 1 KIT2 290,00 €
- 1.16854.0001Superspher® 60 RP-8 endcapped LiChroCART®Superspher® 60 RP-8 endcapped LiChroCART®RP-84 µm4,00 mm125 mmLog in for availability1 * 1 ST883,00 €
- 1.16857.0001Superspher® 60 RP-8 endcapped LiChroCART®Superspher® 60 RP-8 endcapped LiChroCART®RP-84 µm4,00 mm250 mmLog in for availability1 * 1 ST993,00 €
- 1.16052.0001Superspher® 60 RP-8 LiChroCART®
- 1.16052.1003Superspher® 60 RP-8 LiChroCART® (3 columns from 3 batches)Superspher® 60 RP-8 LiChroCART® (3 columns from 3 batches)RP-84 µm4,00 mm125 mmLog in for availability1 * 1 KIT
- 1.16010.0001Superspher® 60 RP-8 LiChroCART®
- 1.16010.1003Superspher® 60 RP-8 LiChroCART® (3 columns from 3 batches)Superspher® 60 RP-8 LiChroCART® (3 columns from 3 batches)RP-84 µm4,00 mm250 mmLog in for availability1 * 1 KIT2 330,00 €
- 1.50791.0001Superspher® 60 RP-select B LiChroCART®Superspher® 60 RP-select B LiChroCART®RP-select B4 µm3,00 mm125 mmLog in for availability1 * 1 ST983,00 €
- 1.50975.0001Superspher® 60 RP-select B LiChroCART®Superspher® 60 RP-select B LiChroCART®RP-select B4 µm4,00 mm125 mmLog in for availability1 * 1 ST1 120,00 €
- 1.50973.0001Superspher® 60 RP-select B LiChroCART®Superspher® 60 RP-select B LiChroCART®RP-select B4 µm4,00 mm250 mmLog in for availability1 * 1 ST1 100,00 €
- 1.50973.1003Superspher® 60 RP-select B LiChroCART® (3 columns from 3 batches)Superspher® 60 RP-select B LiChroCART® (3 columns from 3 batches)RP-select B4 µm4,00 mm250 mmLog in for availability1 * 1 ST
- 1.50974.0001Superspher® 60 RP-select B LiChroCART®Superspher® 60 RP-select B LiChroCART®RP-select B4 µm4,00 mm75 mmLog in for availability1 * 3 ST1 120,00 €
- 1.50198.0001Superspher® RP-18 endcapped LiChroCART®Superspher® RP-18 endcapped LiChroCART®RP-184 µm2,00 mm125 mmLog in for availability1 * 1 ST883,00 €
- 1.50193.0001Superspher® RP-18 endcapped LiChroCART®Superspher® RP-18 endcapped LiChroCART®RP-184 µm2,00 mm250 mmLog in for availability1 * 1 ST853,00 €
- 1.51909.0001Superspher® RP-18 endcapped LiChroCART®Superspher® RP-18 endcapped LiChroCART®RP-184 µm3,00 mm125 mmLog in for availability1 * 1 ST682,00 €
- 1.51910.0001Superspher® RP-18 endcapped LiChroCART®Superspher® RP-18 endcapped LiChroCART®RP-184 µm3,00 mm250 mmLog in for availability1 * 1 ST833,00 €
- 1.16054.0001Superspher® Si 60 LiChroCART®
- 1.16009.0001Superspher® Si 60 LiChroCART®