ContactUsLinkComponent
Thermocouple
Thermocouple
  NABE801200000
Thermocouple
  NABE801200000
  • Description:
    Thermocouple for muffle furnaces, max. 1100 °C
  • Pk:
    1
  • Type:
    Type K
This thermocouple is suitable for muffle furnaces.