ContactUsLinkComponent
Drop test titration kits and reagent packs, MiniKit
Drop test titration kits and reagent packs, MiniKit
Catalog # LOVI56K005501
Supplier:  Lovibond
CAS Number:  
Drop test titration kits and reagent packs, MiniKit
Catalog # LOVI56K005501
Supplier:  Lovibond
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Chlorine (total available) HR drop test kit
  • Range:
    0,2 − 15% Cl2
Specifications
About this item
Derived from laboratory burette methods, the drop test range is an established methodology used across industry and which forms an integral part of the analytical program of many a speciality chemical supplier. The reagents are quick to use and give results fast, making the drop test the method of choice for many customers.
Delivery: Kit is supplied with all reagents and equipment to perform the test, housed in a plastic case. Reagent pack is supplied in a carton containing test reagents and instruction sheet only.