Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Microarray scanner, GenePix® 4100A

Microarray scanner, GenePix® 4100A, configurable, without laser
  MLDVGENEPIX4100A-U
Microarray scanner, GenePix® 4100A
  MLDVGENEPIX4100A-U
Microarray scanner, GenePix® 4100A
Microarray scanner, GenePix® 4100A

Microarray scanner, GenePix® 4100A, configurable, without laser

 
 
   
1 * 1 ST
 

GenePix® 4100A Microarray Scanner offers its simplicity, reliability and flexibility in microarray-based research, be it in genomics, proteomics, or novel applications. It has all the quality, sensitivity, reliability and ease-of use of more expensive scanners, but in a price range and bench top footprint that makes it ideal for individual lab use.

  • Compact, affordable and easy-to-use
  • Superior imaging accuracy
  • Outstanding reproducibility
  • Flexible fluorophore collection
  • Optimise settings on-the-fly  
  • Greater signal-to-noise ratio

The GenePix® 4100A microarray scanner and GenePix® Pro microarray analysis software have been designed to work together as a complete integrated platform. The seamless communication between scanner and software ensures unmatched efficiency for data acquisition and analysis, as well as for real-time scanner performance monitoring. Optional microarray informatics software completes the package, offering database storage, clustering algorithms, advanced statistics and visualisations.

GenePix® Pro acquisition and analysis software automatically reads barcodes from preview scans, data scans, and saved images, and are exported with extracted data.

 : Supplied with 532 nm green and 635 nm red laser, standard filters, USB cable, power cord, calibration slides, 1 year warranty covering parts and labour.

 
  • Description:
    Microarray scanner, GenePix® 4100A, configurable, without laser
  • Seq:
    01
Accessories

 

  •  
    Description
     
     
  • MLDV1-2300-0744
    Green laser kit, 532 nm
       
    1 * 1 ST

 

  •  
    Description
     
     
  • MLDV1-4040-0013
    Red laser kit, 635 nm
       
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.