Anti-CIAS1/NALP3 Polyclonal Antibody
Catalog # BSBTPB9114
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:CIAS1/NALP3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for NALP3/NLRP3 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. NALP3/NLRP3 information: Molecular Weight: 118173 MW; Subcellular Localization: Cytoplasm ; Tissue Specificity: Expressed in blood leukocytes. Strongly expressed in polymorphonuclear cells and osteoblasts. Undetectable or expressed at a lower magnitude in B- and T-lymphoblasts, respectively. High level of expression detected in chondrocytes. Detected in non-keratinizing epithelia of oropharynx, esophagus and ectocervix and in the urothelial layer of the bladder.
Type: Primary
Antigen: CIAS1/NALP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human