Anti-CBS Polyclonal Antibody
Catalog # BSBTRP1041
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:CBS
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for CBS detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CBS information: Molecular Weight: 60587 MW; Subcellular Localization: Cytoplasm . Nucleus ; Tissue Specificity: In the adult strongly expressed in liver and pancreas, some expression in heart and brain, weak expression in lung and kidney. In the fetus, expressed in brain, liver and kidney.
Type: Primary
Antigen: CBS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat