ContactUsLinkComponent
VWR® Hitachi, Spare parts for HPLC system, Chromaster
VWR® Hitachi, Spare parts for HPLC system, Chromaster
Catalog # HITA893-0809
Supplier:  VWR Collection
undefined
VWR® Hitachi, Spare parts for HPLC system, Chromaster
Catalog # HITA893-0809
Supplier:  VWR Collection
Specifications
  • Description:
    Injection valve seal, PEEK, for Chromaster 5210 and 5260 autosamplers
  • Seq:
    575
  • Pk:
    1
Specifications
About this item
This spare part is suitable for Chromaster 5210 and 5260 autosamplers. For Chromaster 5210 AS from Serial No. 1309-001 and 1309T-001 onward. For Chromaster 5260 AS from Serial No. 1205-001 and 1205T-001 onward.