Order Entry
Austria
ContactUsLinkComponent
 
Cryoboxes, PP/PC, B175-series
  GLWAB175B
 :  GLW
undefined
Cryoboxes, PP/PC, B175-series
  GLWAB175B
 :  GLW

 

  • Compartments:
    5×5 (25)
  • W×D×H ext.:
    130×130×70/80 mm
  • Colour:
    Blue
  • Pk:
    1

 

 

These cryo boxes are designed for 5,0 ml test tubes and eppendorf tubes® (5,0 ml).

  • Autoclavable up to +121 °C and use in freezer down to –90 °C
  • More suitable tubes are 5,0 ml E-cups and 5,0 ml micro tubes
  • A clear lid offers a view of the contents without opening the box
  • Drainage holes in base for ease of access at low temperatures

The samples can be perfectly stabilised with and without a label. Ideal for storing, transporting, freezing and autoclaving vials.