Orders LinkContactUsLinkComponent
Multi-channel pipettes, mechanical, variable volume, Reference® 2 (General Lab Product)
Multi-channel pipettes, mechanical, variable volume, Reference® 2 (General Lab Product)
   Sustainable Options Available
Supplier:   EPPENDORF
Multi-channel pipettes, mechanical, variable volume, Reference® 2 (General Lab Product)
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
Multi-channel pipettes, mechanical, variable volume, Reference® 2 (General Lab Product)
Multi-channel pipettes, mechanical, variable volume, Reference® 2 (General Lab Product)
6 Options Available Below
Product Details & Documents
The robust Reference® 2 is the successor to the Reference® pipette and features a new design, reduced weight and unique one-button operation.
  • One-button operation and spring loaded tip cone for fast and ergonomic handling with reduced operating effort
  • Quick tip ejection with active aerosol reduction; haptic feedback prevents accidential tip ejection
  • Spring-loaded tip cone can be switched on/off optionally and improves reproducibility and flexibility
  • Channel indicator guarantees a continuously identical pipette alignment during the work process
  • Serial number is printed on multiple components of the pipette for heightened traceability
  • Embedded RFID chip containing all relevant data ensures complete traceability
  • Optional TrackIT software allows simple identification and documentation and provides ability to add own data (e.g. calibration time and results, location)
  • Fully autoclavable
Thanks to its reliable and accurate operation, the pipette is ideal for use with precious liquids or in any application requiring extremely high levels of accuracy.
Designed and constructed for general laboratory and research applications in Life Science. Typical applications include: Food & beverages, forensic, cosmetics, agriculture, animal studies, chemical analytics, research applications at medical and academic facilities.
* Works with Windows 7/8 and Vista.
Certifications: Without CE marking.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-5843
  • 613-5845
  • 613-5847
  • 613-5844
  • 613-5846
  • 613-5848
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifications
Product Family Options
8-channel
Product Information
  • Volume
    Accuracy
    Imprecision
    Colour code
    Recommended tips
    Availability
    Price
  • 0,5 - 10 µl
    0,5 µl: ±12,0 % / ±0,06 µl
    1 µl: ±8,0 % / ±0,08 µl
    5 µl: ±4,0 % / ±0,2 µl
    10 µl: ±2,0 % / ±0,2 µl
    0,5 µl: ±8,0 % / ±0,04 µl
    1 µl: ±5,0 % / ±0,05 µl
    5 µl: ±2,0 % / ±0,1 µl
    10 µl: ±1,0 % / ±0,1 µl
    Grey
    20 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 037,00 €
    1 037,00 €
    1 * 1 ST
       1 037,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5843
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 10 - 100
    10 µl: ±3,0 % / ±0,3 µl
    50 µl: ±1,0 % / ±0,5 µl
    100 µl: ±0,8 % / ±0,8 µl
    10 µl: ±2,0 % / ±0,2 µl
    50 µl: ±0,8 % / ±0,4 µl
    100 µl: ±0,3 % / ±0,3 µl
    Yellow
    200 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 037,00 €
    1 037,00 €
    1 * 1 ST
       1 037,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5845
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 30 - 300 µl
    30 µl: ±3,0 % / ±0,9 µl
    150 µl: ±1,0 % / ±1,5 µl
    300 µl: ±0,6 % / ±1,8 µl
    30 µl: ±1,0 % / ±0,3 µl
    150 µl: ±0,5 % / ±0,75 µl
    300 µl: ±0,3 % / ±0,9 µl
    Orange
    300 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 037,00 €
    1 037,00 €
    1 * 1 ST
       1 037,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5847
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
12-channel
Product Information
  • Volume
    Accuracy
    Imprecision
    Colour code
    Recommended tips
    Availability
    Price
  • 0,5 - 10 µl
    0,5 µl: ±12,0 % / ±0,06 µl
    1 µl: ±8,0 % / ±0,08 µl
    5 µl: ±4,0 % / ±0,2 µl
    10 µl: ±2,0 % / ±0,2 µl
    0,5 µl: ±8,0 % / ±0,04 µl
    1 µl: ±5,0 % / ±0,05 µl
    5 µl: ±2,0 % / ±0,1 µl
    10 µl: ±1,0 % / ±0,1 µl
    Grey
    20 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 135,00 €
    1 135,00 €
    1 * 1 ST
       1 135,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5844
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 10 - 100 µl
    10 µl: ±3,0 % / ±0,3 µl
    50 µl: ±1,0 % / ±0,5 µl
    100 µl: ±0,8 % / ±0,8 µl
    10 µl: ±2,0 % / ±0,2 µl
    50 µl: ±0,8 % / ±0,4 µl
    100 µl: ±0,3 % / ±0,3 µl
    Yellow
    200 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 135,00 €
    1 135,00 €
    1 * 1 ST
       1 135,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5846
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 30 - 300 µl
    30 µl: ±3,0 % / ±0,9 µl
    150 µl: ±1,0 % / ±1,5 µl
    300 µl: ±0,6 % / ±1,8 µl
    30 µl: ±1,0 % / ±0,3 µl
    150 µl: ±0,5 % / ±0,75 µl
    300 µl: ±0,3 % / ±0,9 µl
    Orange
    300 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 135,00 €
    1 135,00 €
    1 * 1 ST
       1 135,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5848
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Pipette holder for pipette carousel
    Pipette holder for pipette carousel
    613-2925
    Log in for availability
    1 * 1 ST - 19,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Protection filter for 0,25 - 2,5 ml variable / 2 ml fixed volume / 2,5 ml fixed volume, colour code red, 1 set = 10 filters, 1 sleeve
    Protection filter for 0,25 - 2,5 ml variable / 2 ml fixed volume / 2,5 ml fixed volume, colour code red, 1 set = 10 filters, 1 sleeve
    EPPE4920622.009
    Log in for availability
    1 * 1 ST - 17,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Protection filter for 1 - 10 ml variable, colour code turquoise, 1 set = 10 filters, 1 sleeve
    Protection filter for 1 - 10 ml variable, colour code turquoise, 1 set = 10 filters, 1 sleeve
    EPPE4920624.001
    Log in for availability
    1 * 1 ST - 17,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Protection filter for 0,5 - 5 ml variable, colour code purple, 1 set = 10 filters, 1 sleeve
    Protection filter for 0,5 - 5 ml variable, colour code purple, 1 set = 10 filters, 1 sleeve
    EPPE4920623.005
    Log in for availability
    1 * 1 ST - 17,00 €