
Ganciclovir 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:82410-32-0
:
Formula:C₉H₁₃N₅O₄
MDL Number:MFCD00870588
:255.23 g/mol
Melting Pt:251…252 °C
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤4 %
- HPLC:≥97.5 %
Specification Test Results
Related Information
Frequently Bought Together