Irinotecan hydrochloride trihydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:136572-09-3
:MFCD01862255
Formula:C₃₃H₃₈N₄O₆·HCl·3H₂O
MDL Number:MFCD01765731
:677.19 g/mol (623.15 anhy)
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Water:7 to 9 %
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together