Order Entry
Austria
ContactUsLinkComponent
 
Triethylamine for HPLC
Catalog # 460812500.
CAS Number:  
undefined
Triethylamine for HPLC
Catalog # 460812500.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 %
  • GC:
    ≥99.4 %
  • UV:
    at 254 nm A: ≤0.125 (0.1 M in water)
  • Residue after evaporation:
    ≤0.01 %
  • Iron (Fe):
    ≤0.2 ppm
  • Lead (Pb):
    ≤0.02 ppm

Specification Test Results

About this item

Synonyms: (Diethylamino)ethane, TEA
 :
 :101.19 g/mol
UN:1296
 :GHS06
 :GHS05
 :GHS02
Melting Pt:–115 °C
Storage Temperature:Ambient
CAS Number:121-44-8
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051
Boiling Pt:90 °C (760 mmHg)
Density:0.728 g/cm³
Flash Pt:–11 °C
ADR:3 II
Merck Index:13,09740