Order Entry
Austria
ContactUsLinkComponent
Ammonia solution 35% in water for HPLC
 
 
 
Ammonia solution 35% in water for HPLC
  ACRO460801000
 :  
undefined
 
 
 
Ammonia solution 35% in water for HPLC
  ACRO460801000
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Appearance:
    Clear colorless Solution
  • UV:
    at 254 nm A: ≤0.01
  • Titration with HCl:
    32 to 36 %
  • Iron (Fe):
    ≤0.2 ppm
  • Lead (Pb):
    ≤0.05 ppm

 

 

 :
 :35.05 g/mol
 :0.88 g/cm³
 :Ambient
 :2672
 :8,III
 :GHS05
 :GHS07
 :GHS09
 :1336-21-6
 :7664-41-7
 :NH₃
 :MFCD00066650