Order Entry
Austria
ContactUsLinkComponent
Copper (II) ethylenediamine 1 M in water, Technical Grade not for viscosity determination
Corrosive
Copper (II) ethylenediamine 1 M in water, Technical Grade not for viscosity determination
Catalog # ACRO460680010
CAS Number:  
undefined
Corrosive
Copper (II) ethylenediamine 1 M in water, Technical Grade not for viscosity determination
Catalog # ACRO460680010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Dark blue Solution
  • Strength:
    0.98 to 1.02 M (copper molarity)

Specification Test Results

About this item

 :
 :217.76 g/mol
Density:1.1 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:1761
ADR:8,II
 :GHS05
CAS Number:14552-35-3
 :13426-91-0
Formula:Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MDL Number:MFCD00274628