Order Entry
Austria
ContactUsLinkComponent
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8) 98%
Irritant
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8) 98%
Certificates
undefined
Irritant
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8) 98%

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 %
  • Solubility:
    (10% in water) Clear solution
  • HPLC:
    ≥97.5 %

Specification Test Results

Frequently Bought Together