Bile, bovine, powder
- CAS: 8008-63-7
- Solubility:Soluble in water
- Physical form:powder
Frequently Bought Together
NBB®-B, Broth
: DOEHLER GROUP
VWR® Dip Slides
: VWR Chemicals
VWR Chemicals, Base Materials for Dehydrated Media
: VWR Chemicals
Oxidase, Bactident®, Millipore®
: Merck
Bioindicator, Sterikon® plus
: Merck
Environmental surface sampling bags, Whirl-Pak® Speci-Sponge®
: NASCO FORT ATKINSON
VWR® Dehydrated Culture Media, Powdered
: VWR Chemicals
Dehydrated culture media
: Merck
Accessories
: Merck
VWR® Culture Media, Ready to Use, in Bottles
: VWR Chemicals
VWR® Culture Media, Ready To Use, In Plates
: VWR Chemicals
Accessories for Anaerobic Systems, Oxoid
: THERMO FISHER DIAGNOSTICS
VWR® Contact Plates, Blister Packed, Irradiated
: VWR Chemicals