Order Entry
Austria
ContactUsLinkComponent
Ammonia solution 10% (w/w) in water, extra pure
Corrosive
Irritant
Ammonia solution 10% (w/w) in water, extra pure
Certificates
undefined
Corrosive
Irritant
Ammonia solution 10% (w/w) in water, extra pure

Specification Test Results

  • Appearance:
    Clear solution
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.002 %
  • Strength:
    9.5 to 10.5 %

Specification Test Results

Frequently Bought Together