Order Entry
Austria
ContactUsLinkComponent
Moxifloxacin hydrochloride
Moxifloxacin hydrochloride
Catalog # ACRO457960010
CAS Number:  
undefined
Moxifloxacin hydrochloride
Catalog # ACRO457960010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Off-white to yellow Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤5 %
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

Synonyms: 1-Cyclopropyl-6-fluoro-7-((4aS,7aS)-hexahydro-1H-pyrrolo[3,4-b]pyridin-6(2H)-yl)-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, Moxifloxacin HCl
 :
 :437.896 g/mol
Melting Pt:324…325 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:186826-86-8
Formula:C₂₁H₂₅ClFN₃O₄
MDL Number:MFCD00949117