ContactUsLinkComponent
Iodine, meets analytical specification of BP, Ph. Eur., USP
Iodine, meets analytical specification of BP, Ph. Eur., USP
Certificates
undefined
Iodine, meets analytical specification of BP, Ph. Eur., USP
Certificates
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Greyish-violet Brittle plates or crystals with metallic sheen
  • Titration Iodometric:
    99.8 to 100.5 %
  • Non-volatile matter:
    ≤0.05 %
  • Chloride and Bromide:
    ≤250 ppm