Acetic acid 25% in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
Boiling Pt:103 °C (760 mmHg)
Melting Pt:–17 °C
Density:1.04 g/cm³
Molecular Weight:60.05 g/mol
Flash Pt:>100 °C
Storage Temperature:Ambient
UN:2790
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Titration with NaOH:24.0 to 26.0 %
Specification Test Results
Frequently Bought Together