Acetone, meets analytical specification of Ph.Eur.
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Boiling Pt:56 °C (760 mmHg)
Melting Pt:–95 °C
Density:0.79 g/cm³
Flash Pt:–20 °C
Storage Temperature:Ambient
UN:1090
ADR:3,II
:GHS07
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.3 %
- Appearance of solution:Clear colorless
- Identification:Passes test
- Relative density:0.790 to 0.793
- Residue after evaporation:≤50 ppm
- Related substances:≤0.05 % (Methanol)
- Acidity/alkalinity:Passes test
- Insoluble matter:(in water) Passes test
- Reducing substances:Passes test
Specification Test Results
Frequently Bought Together