Tobramycin 95+%
:32986-56-4
:
:C₁₈H₃₇N₅O₉
:MFCD00077885
:467.51 g/mol
:Raumtemperatur
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Water:≤8 %
- Titration with HClO4:≥95.0 % (on dry substance)
Frequently Bought Together