Order Entry
Austria
ContactUsLinkComponent
Calcium hydroxide 99.995% (trace metals basis)
Corrosive
Irritant
Calcium hydroxide 99.995% (trace metals basis)
Certificates
undefined
Corrosive
Irritant
Calcium hydroxide 99.995% (trace metals basis)
Certificates

Specification Test Results

  • Appearance:
    White Powder or crystals
  • Titration Complexometric:
    ≥54 % (Ca)
  • Total trace metal impurities:
    ≤50 ppm

Specification Test Results

Related Information

Frequently Bought Together