2-Cyclohepten-1-one 90%
:1121-66-0
:
:C₇H₁₀O
:MFCD00004157
:110.16 g/mol
:189 °C (760 mmHg)
:0.99 g/cm³
:68 °C
:GHS07
:Ambient
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥89.0 %
- HPLC:≥94 %
Frequently Bought Together