Order Entry
Austria
ContactUsLinkComponent
Tetrahydrouridine ≥90% (by TLC), Millipore®
Tetrahydrouridine ≥90% (by TLC), Millipore®
Catalog # 584222-10
CAS Number:  
Tetrahydrouridine ≥90% (by TLC), Millipore®
Catalog # 584222-10
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :248.2331 g/mol
Storage Temperature:Freezer
CAS Number:18771-50-1
Formula:C₉H₁₆N₂O₆
MDL Number:MFCD00063468