Chemical resistant overalls, 9877
Supplier: UVEX
Certificates
About this item
These disposable overalls are made of a very light, microporous PE laminate, combined with an SMS back section for high breathability. The overalls guarantee a high level of protection against liquid splashes and solid particles.
- Particle-tight and spray-tight to a limited extent
- Free of silicone and paint wetting impairment substances
- Bound seams in contrasting colours for visible safety
- Middle finger loops prevent sleeves sliding up the arm
- Self-adhesive zipper flap for optimal protection
- Optimal fit with elasticated waistband and elasticated bands on hood, arms and legs
- Anti-static properties
Typical applications include: Handling chemicals in dust and powder form, varnishing and protection against paint splashes, fibreglass production and processing, industrial cleaning and maintenance, automotive industry, grinding and polishing, working with asbestos, wood and metal processing, construction industry, cleanroom applications, pharmaceutical industry.
Suitable for cleanroom environments Class 8 in accordance with ISO 14644-1.
Certifications: Type 5/6, EN 1149-5; Cat. III
UV protection factor UPF 50+
UV protection factor UPF 50+
Specifications
- Catalog No:
- 111-2614
- UVEX9871014
- UVEX9871010
- UVEX9871011
- UVEX9871013
- Model:
- 9877
- 9877
- 9877
- 9877
- 9877
- Anti-static:
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial